CleanCap® AU EGFP saRNA
Lieferant: TriLink BioTechnologies
L-7801-100 L-7801-1000 L-7801-5 L-7901-100 L-7901-1000 L-7901-5
SOLUL-7801-100EA 388 EUR
SOLUL-7801-100 SOLUL-7801-1000 SOLUL-7801-5 SOLUL-7901-100 SOLUL-7901-1000 SOLUL-7901-5
CleanCap® AU EGFP saRNA
Protein- Expression Proteinexpressions-Reagenzien
This construct expresses an enhanced version of the green fluorescent protein (EGFP), originally isolated from the jellyfish Aequorea victoria, as self-amplifying RNA (saRNA). EGFP is a commonly used direct detection reporter in mammalian cells, yielding bright green fluorescence with an emission peak at 509 nm.
  • Due to their self-amplification capability, saRNAs may provide more durable and longer protein expression than mRNAs. This ready-to-use saRNA construct is:
  • Capped using CleanCap® Reagent AU, mimicking the authentic 5′ alphavirus end while providing an optimal Cap-1 structure with >95% capping efficiency, for efficient expression in mammalian cells.
  • Synthesized using CleanScribe™ RNA Polymerase, reducing unwanted double-stranded RNA (dsRNA) by-products by up to 85%, for lower inflammatory responses and enhanced functionality.
  • Manufactured using TriLink’s proprietary CleanScript® saRNA in vitro transcription (IVT) method, improving saRNA yield and integrity while also lowering dsRNA formation, for enhanced protein expression.
  • Unmodifid or modified with 5-methylcytidine (5mC), which can increase nuclease stability, reduce interaction of innate immune receptors, and enhance translational efficiency of saRNAs.
Our catalog saRNA construct employs alphaviral replication machinery from Venezuelan equine encephalitis virus (VEEV) that encodes for the non-structural proteins (nsP1-4). It is polyadenylated (26A) and optimized for mammalian systems.
Concentration: 1 mg/ml