TriLink® N1-Ethylpseudouridine-5'-Triphosphate
Lieferant: TriLink BioTechnologies
A modified NTP for incorporation into messenger RNAs (mRNA) using T7 RNA Polymerase. Incorporation of N1-ethylpseudouridine can reduce the immunogenicity of the resulting mRNA.
- Grade: RUO
Bitte beachten Item will be delivered at or below –20 °C and should be stored at or below –20 °C.