TriLink® N1-Ethylpseudouridine-5'-Triphosphate
Lieferant: TriLink BioTechnologies
N-1102-1
N-1102-10
N-1102-100
N-1102-5
SOLUN-1102-1EA
75.3
EUR
SOLUN-1102-1
SOLUN-1102-10
SOLUN-1102-100
SOLUN-1102-5
TriLink® N1-Ethylpseudouridine-5'-Triphosphate
Nukleinsäure-Reagenzien
Nukleotide
A modified NTP for incorporation into messenger RNAs (mRNA) using T7 RNA Polymerase. Incorporation of N1-ethylpseudouridine can reduce the immunogenicity of the resulting mRNA.
- Grade: RUO
Bitte beachten Item will be delivered at or below –20 °C and should be stored at or below –20 °C.