TriLink® N1-Ethylpseudouridine-5'-Triphosphate
Lieferant: TriLink BioTechnologies
N-1102-1 N-1102-10 N-1102-100 N-1102-5
SOLUN-1102-1EA 75.3 EUR
SOLUN-1102-1 SOLUN-1102-10 SOLUN-1102-100 SOLUN-1102-5
TriLink® N1-Ethylpseudouridine-5'-Triphosphate
Nukleinsäure-Reagenzien Nukleotide
A modified NTP for incorporation into messenger RNAs (mRNA) using T7 RNA Polymerase. Incorporation of N1-ethylpseudouridine can reduce the immunogenicity of the resulting mRNA.
  • Grade: RUO
Bitte beachten Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78b41472f9125d8c1706bd623e88b5df69852a26&vid=78b41472f9125d8c1706bd623e88b5df69852a26&prid=web&clid=SAPDAM