TriLink® Uridine-5'-Triphosphate
Lieferant: TriLink BioTechnologies
N-1504-10
N-1504-100
N-1504-25
SOLUN-1504-10EA
25.3
EUR
SOLUN-1504-10
SOLUN-1504-100
SOLUN-1504-25
TriLink® Uridine-5'-Triphosphate
Nukleinsäure-Reagenzien
Nukleotide
This nucleotide consists of an unmodified base, a ribose, and a 5′ triphosphate group for synthesis of RNA molecules.
- Unmodified ribonucleotide for RNA synthesis
- >99% purity by AX-HPLC analysis
- Provided as 100 mM in H₂O, pH 7,5
- Grade: RUO
Bitte beachten Item will be delivered at or below –20 °C and should be stored at or below –20 °C.