TriLink® N7-Methyl-3'-O-Methyl-Guanosine-5'-Triphosphate-5'-Guanosine
Lieferant: TriLink BioTechnologies
N-7003-1
SOLUN-7003-1EA 143 EUR
SOLUN-7003-1
TriLink® N7-Methyl-3'-O-Methyl-Guanosine-5'-Triphosphate-5'-Guanosine
Nukleinsäure-Reagenzien Nukleotide
  • Grade: RUO
Trusted TriLink quality, exceptional price. A key step in cellular mRNA processing is the addition of a 5’ cap structure, which is a 5'-5' triphosphate linkage between the 5' end of the RNA and a guanosine nucleotide. The cap is methylated enzymatically at the N-7 position of the guanosine to form mature mCAP. When preparing synthetic mRNA, the cap is often added prior to use in order to stabilise the mRNA and significantly enhance translation. Using a 4:1 mixture of a cap analog to GTP in transcription reactions will cap 80% of the resulting mRNAs. If mCAP (pre-methylated CAP) is used, only half of the asymmetrical mCAP will insert in the functional orientation. ARCA can only insert in the correct orientation, resulting in mRNAs that are translated twice as efficiently. TriLink is the least expensive source of high quality ARCA. We also offer CAP and mCAP for more traditional capping reactions.
Bitte beachten Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b79dd382bb9cb2010337f6b7b3def6e6e7c2e5c&vid=3b79dd382bb9cb2010337f6b7b3def6e6e7c2e5c&prid=web&clid=SAPDAM