TriLink® 2',3'-Dideoxyadenosine-5'-O-(1-Thiotriphosphate)
Lieferant: TriLink BioTechnologies
N-8009-1 N-8009-10 N-8009-5
SOLUN-8009-1EA 94.3 EUR
SOLUN-8009-1 SOLUN-8009-10 SOLUN-8009-5
TriLink® 2',3'-Dideoxyadenosine-5'-O-(1-Thiotriphosphate)
Nukleinsäure-Reagenzien Nukleotide
2',3'-Dideoxyadenosine-5'-O-(1-Thiotriphosphate) is a sugar modified nucleoside triphosphate where the 2' and 3' hydroxyl groups are absent, resulting in chain termination. This ddATP analog also contains a 1-Thiotriphosphate which results in a nuclease resistant phosphorothioate linkage. The inability of polymerases to extend from a dideoxy nucleotide causes chain termination and is useful in antiviral research and in a variety of biotechnology applications.
  • Grade: RUO
Bitte beachten Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb2a9a0b0d1574d7e19699e3b023429780985971&vid=bb2a9a0b0d1574d7e19699e3b023429780985971&prid=web&clid=SAPDAM