Flashpoint tester
Lieferant: STANHOPE SETA
13811-3
STAN13811-3EA 0 EUR
STAN13811-3
Flashpoint tester
Flammpunktprüfer
Semi-automatic tester equipped with an electrically heated cup, a button-operated and electrically driven sweep arm and a test flame used for industrial applications with greases.
  • Variable manually controlled ramp rate
  • Easy to clean stainless steel
  • Variable control to set temperature rise rate
Standards: AASHTO T48, D92, BS 2000-36 , EN ISO 2592, FTM 791 1103.7, IP 36.
Lieferumfang: Supplied with instruction manual and mains lead.