StemSpan™ CC100 Culture Supplement
Lieferant: STEMCELL Technologies
02690
STMC02690EA 2070 EUR
STMC02690
StemSpan™ CC100 Culture Supplement
Zellkulturmedien
Serum-free culture supplement for expansion of human hematopoietic cells.
StemSpan™ CC100 cytokine cocktail contains a combination of both early- and late-acting recombinant human (rh) cytokines formulated to support the proliferation of human hematopoietic cells. It is supplied as a 100X concentrate. When added to serum-free medium, StemSpan™ CC100 promotes the expansion of CD34+ cells isolated from human cord blood and bone marrow, and is recommended if high cell yields and large numbers of CD34+ cells are desired.
We recommend using StemSpan™ CC100 in combination with any of the following StemSpan™ media:
StemSpan™ SFEM
StemSpan™ SFEM II
StemSpan™-XF
StemSpan™-AOF
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').