ES-Cult™ M3120 Medium
Lieferant: STEMCELL Technologies
03120
STMC03120EA 215 EUR
STMC03120
ES-Cult™ M3120 Medium
Zellkulturmedien
Base methylcellulose medium for in vitro differentiation of mouse ES or iPS cells to hematopoietic or endothelial progenitor cells.
ES-Cult™ M3120 is recommended as a base for the preparation of methylcellulose-based medium for the in vitro differentiation of mouse embryonic stem (ES) or induced pluripotent stem (iPS) cells into hematopoietic or endothelial progenitor cells. When supplemented with the appropriate additional components, ES-Cult™ M3120 can be used for the generation of embryoid bodies (EBs) from undifferentiated mouse ES or iPS cells.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').