MethoCult™ SF M3236 Medium
Lieferant: STEMCELL Technologies
03236
STMC03236EA 577 EUR
STMC03236
MethoCult™ SF M3236 Medium
Zellkulturmedien
Serum-free methylcellulose-based medium without cytokines for mouse cells.
MethoCult™ SF M3236 is a serum-free, incomplete medium formulation that contains methylcellulose in Iscove's MDM. MethoCult™ SF M3236 is suitable for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of mouse bone marrow, spleen, and fetal liver, when the appropriate growth factors and supplements are added. This formulation does not contain serum or cytokines and is ideal for testing cytokine effects where the presence of fetal bovine serum is not desired.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').