ClonaCell™ CHO ACF Supplement
Lieferant: STEMCELL Technologies
03820
STMC03820EA 254 EUR
STMC03820
ClonaCell™ CHO ACF Supplement
Zellkulturmedien
Animal component-free, serum-free, glutamine-free 40X supplement for cloning during CHO cell line development.
ClonaCell™-CHO ACF supplement is a serum-free and animal component-free supplement that is added to protein-free media in order to increase cloning efficiency and promote robust growth of CHO cells. The 40X supplement can be added to a variety of protein-free liquid or semi-solid cell culture media to increase the growth and single-cell cloning efficiency of suspension-adapted CHO cells. The supplement contains recombinant proteins and phenol red, and does not contain animal components, serum, hydrolysates, L-glutamine, or selection agents.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').