ClonaCell™-HY Liquid HAT Selection Medium
Lieferant: STEMCELL Technologies
03831
STMC03831EA 574 EUR
STMC03831
ClonaCell™-HY Liquid HAT Selection Medium
Zellkulturmedien
Hybridoma selection medium, with HAT (serum-containing).
ClonaCell™-HY liquid HAT selection medium is a serum-containing medium with hypoxanthine, aminopterin, and thymidine (HAT) for the selection of hybridomas following the fusion of lymphocytes and myeloma cells. This medium has been verified for use in mouse and rat hydridoma development and reportedly is compatible for production and cloning of myelomas and/or hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').