mTeSR™ 3D Medium
Lieferant: STEMCELL Technologies
03950
STMC03950EA 1010 EUR
STMC03950
mTeSR™ 3D Medium
Zellkulturmedien
Serum-free media for suspension culture of human ES and iPS cells.
mTeSR™3D was developed for expansion and scale-up of undifferentiated human embryonic stem (ES) cells and human induced pluripotent stem (iPS) cells as aggregates in 3D suspension culture. The novel fed-batch workflow saves time and media. Daily feeds replenish nutrients and eliminate the need for medium exchanges on non-passaging days. mTeSR™3D is a specialised and serum-free formulation. This culture system is compatible with a range of suspension culture vessels.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').