MethoCult™ H4531 Medium
Lieferant: STEMCELL Technologies
04531
STMC04531EA 1080 EUR
STMC04531
MethoCult™ H4531 Medium
Zellkulturmedien
Methylcellulose-based medium with agar-LCM (without EPO) for human cells.
MethoCult™ H4531 is methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, peripheral blood, mobilised peripheral blood, and cord blood samples. This formulation contains agar leukocyte conditioned medium (agar-LCM) as a source of colony-stimulating factors. MethoCult™ H4531 supports the growth of granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M) and EPO-independent erythroid progenitor cells. It is suitable for detection of EPO-independent erythroid progenitor cells.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').