MethoCult™ SF H4536 Medium
Lieferant: STEMCELL Technologies
04536
STMC04536EA 1440 EUR
STMC04536
MethoCult™ SF H4536 Medium
Zellkulturmedien
Serum-free methylcellulose-based medium with recombinant cytokines (without EPO) for human cells.
MethoCult™ SF H4536 is a complete serum-free methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilised peripheral blood, peripheral blood, and cord blood samples. MethoCult™ SF H4536 is formulated to support the optimal growth of granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M) under defined serum-free conditions.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').