STEMdiff™ Hematopoietic Kit
Lieferant: STEMCELL Technologies
05310
STMC05310EA 1140 EUR
STMC05310
STEMdiff™ Hematopoietic Kit
Zellkulturmedien
For differentiation of human PSCs into the hematopoietic lineage.
Generate functional hematopoietic progenitor cells from human embryonic stem (ES) and induced pluripotent stem (iPS) cells with STEMdiff™ Hematopoietic Kit. Using a simple and reproducible protocol in serum- and feeder-free conditions, you can produce hematopoietic progenitor cells expressing CD34, CD45, and CD43. The robust, two-stage protocol initially induces cells toward the mesoderm before further differentiating them into hematopoietic progenitor cells. After 12 days, a population of hematopoietic cells containing 25 to 65% (average 43%) CD34+CD45+ progenitor cells can be harvested, including progenitor cells that have the capacity to form functional hematopoietic colonies in the colony-forming unit (CFU) assay.
STEMdiff™ hematopoietic kit has been optimized for differentiation of human pluripotent cells maintained in:
mTeSR™1
mTeSR™ plus
TeSR™-E8™
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').