MesenCult™ Osteogenic Stimulatory Kit (Mouse)
Lieferant: STEMCELL Technologies
Complete medium for differentiating mouse mesenchymal stem cells and embryonic fibroblasts into osteoblasts.
MesenCult™ osteogenic stimulatory kit (Mouse) is specifically formulated for the in vitro differentiation of mouse mesenchymal stem and progenitor cells (MSCs) from compact bone and bone marrow, and mouse embryonic fibroblasts (MEFs) into osteoblasts. This kit induces strong osteogenesis of mouse MSCs and MEFs, as evidenced by qPCR analysis of key transcripts involved in bone differentiation and maturation, and alizarin red staining. This kit is recommended for characterizing MSCs and MEFs and studying bone development.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').