NeuroCult™ Basal Medium (Mouse and Rat)
Lieferant: STEMCELL Technologies
05700
STMC05700EA 349 EUR
STMC05700
NeuroCult™ Basal Medium (Mouse and Rat)
Zellkulturmedien
Basal medium for culture of mouse and rat neural stem and progenitor cells.
NeuroCult™ basal medium (Mouse and Rat) is a standardised serum-free basal medium designed to be supplemented with NeuroCult™ proliferation supplement (Mouse and Rat) and appropriate cytokines for the in vitro culture and expansion of mouse and rat neural stem and progenitor cells. NeuroCult™ basal medium (Mouse and Rat) can also be supplemented with NeuroCult™ differentiation supplement (Mouse and Rat) for the differentiation of mouse and rat neural stem and progenitor cells into neurons, astrocytes, and oligodendrocytes. NeuroCult™ basal medium (Mouse and Rat) is a component of NeuroCult™ proliferation kit (Mouse and Rat) and NeuroCult™ differentiation kit (Mouse and Rat).
NOTE: When preparing complete NeuroCult™ proliferation medium, addition of human recombinant EGF is required. When culturing cells obtained from adult mouse or rat, human recombinant bFGF and heparin solution are also required.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').