STEMdiff™ Neural Rosette Selection Reagent
Lieferant: STEMCELL Technologies
05832
STMC05832EA 85.7 EUR
STMC05832
STEMdiff™ Neural Rosette Selection Reagent
Zellkultur-Ergänzungen und -Zusätze
Enzyme-free reagent for the selective detachment of neural rosettes.
  • Isolate CNS-type neural progenitor cells rapidly and efficiently, without harsh enzymatic treatment
  • Selectively detach neural rosette clusters without manual scraping
  • Obtain highly pure neural progenitor cell poplulations
Rapidly and efficiently isolate neural rosettes, without harsh enzymatic treatment, with STEMdiff™ neural rosette selection reagent. This enzyme-free reagent enables selective detachment of neural rosette clusters from adherent neural aggregates previously generated from human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells using STEMdiff™ neural induction medium, without manual scraping. Collecting and re-plating rosette clusters after incubation with this reagent will yield highly pure populations of neural progenitor cells (NPCs), which can be further sub-cultured as single cells.
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').