Methylenblau in 3% Essigsäure
Lieferant: STEMCELL Technologies
Synonyme:
3,7-Bis(dimethylamino)phenothiaziniumchlorid, N,N,N′,N′-Tetramethylthioninchlorid, Methylthioniniumchlorid
3% Acetic Acid with Methylene Blue. Reagent for manual counting of nucleated mammalian cells
3% Acetic Acid with Methylene Blue is recommended for manual counting of nucleated mammalian cells. It will lyse red blood cell and white blood cell membranes. The remaining white blood cell nuclei will stain lightly with methylene blue.
Contains
Acetic acid (3%)
Methylene blue
Distilled water
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Formel:
C₁₆H₁₈ClN₃S |
MDL:
MFCD00150008 CAS-Nummer: 61-73-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...