For depletion of red blood cells from fresh blood samples and isolation of nucleated cells.
HetaSep™ is an erythrocyte aggregation agent used to quickly separate nucleated cells from red blood cells (RBCs) in whole blood. Aggregated erythrocytes settle much faster than dispersed cells. By controlling the settling time and/or centrifugation speed, the majority of nucleated cells are recovered in the supernatant. Approximately 95 to 99% RBC depletion is attained if the nucleated cell-rich fraction is removed carefully. HetaSep™ contains 6% w/v hetastarch.
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).