Dispase (5 U/ml)
Lieferant: STEMCELL Technologies
5 U/ml dispase in Hanks' balanced salt solution.
- Achieve gentle dissociation in a wide variety of tissues
- Optimised for for the generation of single-cell suspensions from dissociated human and mouse mammary tissue
Use dispase for gentle dissociation of a wide variety of tissues. Incubation of minced tissue with pre-warmed Dispase and gentle agitation will liberate cells with minimal cell damage. Pre-warmed dispase can also be used to harvest cells from tissue culture plastic. This proteolytic dissociation reagent has been optimized for use in the generation of a single-cell suspension from dissociated human and mouse mammary tissue.
This product contains 5 U/ml Dispase II (neutral protease from Bacillus polymyxa) dissolved in Hanks’ balanced salt solution. Unlike trypsin, Dispase is not inhibited by serum. Dispase activity is inhibited by EDTA and EGTA. Dispase should be removed from cell suspensions by centrifugation followed by washing the cells with buffer or culture medium.
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').