EasySep™ Release Human CD45 Positive Selection Kits
Lieferant: STEMCELL Technologies
100-0105 100-0107 100-0108 100-0109
STMC100-0105EA 773 EUR
STMC100-0105 STMC100-0107 STMC100-0108 STMC100-0109
EasySep™ Release Human CD45 Positive Selection Kits
Zelltrennmedien
Immunomagnetic positive selection of particle-free CD45+ leukocytes from primary human tissues and tumors.
  • Highly purified CD45+ cells isolated in less than 45 minutes
  • No-wash removal of EasySep™ Releasable RapidSpheres™
The EasySep™ Release Human CD45 Positive Selection Kit is designed to isolate CD45+ leukocytes from single-cell suspensions of primary human tissues and tumors, or tissues and tumor xenografts from humanised mice, by immunomagnetic positive selection.
Desired cells are labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Then, bound magnetic particles are removed from the EasySep™-isolated CD45+ cells, which are immediately available for downstream applications. Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands.
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).