EBV (BZLF1) Peptide Pool
Lieferant: STEMCELL Technologies
The EBV (BZLF1) peptide pool is a lyophilised mixture of 59 peptides from trans-activator protein BZLF1 of Epstein-Barr virus (EBV; strain B95-8). BZLF1 is a DNA-binding protein that acts as a switch from latent infection to lytic infection. The pool consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 245 on BZLF1.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').