Cell Navigator™ Golgi Staining Kit, Green
Lieferant: STEMCELL Technologies
Kit containing C6 NBD ceramide for selectively staining the Golgi apparatus in live or aldehyde-fixed cells - 100 tests.
Cell Navigator™ Golgi Staining Kit is an optimized assay kit for staining morphological features of the Golgi apparatus in live and fixed cells. This kit contains NBD ceramide, a fluorescent analog of ceramide-a sphingosine derivative that selectively stains the Golgi apparatus. NBD ceramide fluorescence is weak in water, and increases in aprotic solvents and non-polar solvents. Therefore, NBD ceramide is complexed with bovine serum albumin (BSA) prior to administration into cells; the resulting fluorescent metabolites stain the Golgi apparatus. The kit also contains a staining buffer, DMSO, and Hoechst 33342 - a dye that selectively labels DNA which can be used as a nuclear co-stain.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').