FITC-C6-DEVD-FMK Durchflusscytometrie
Lieferant: STEMCELL Technologies
For detection of active caspase-3 in mammalian cells undergoing apoptosis.
Caspase-3 is a proteolytic enzyme with a substrate selectivity for the peptide sequence Asp-Glu-Val-Asp (DEVD). FITC-C6-DEVD-FMK is a non-toxic, cell-permeable fluorescent substrate that irreversibly binds to activated caspase-3 in apoptotic cells. The FITC-labeled substrate can be detected with high sensitivity using flow cytometry.
Caspases are a family of proteases that play a central role in cellular apoptosis through the cleavage of select proteins, and result in the disassembly of the cell (Thornberry & Lazebnik Science, 1998). This process is switched on by pro-apoptotic signals from death receptors or stimuli such as cytotoxic reagents, which activate the initiator caspases and lead to activation of the effector caspases—caspase-3 and caspase-7 (Thornberry & Lazebnik Science, 1998). FITC-C6-DEVD-FMK is useful for screening caspase-3 inhibitors and quantifying apoptotic cells containing active caspase-3.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').