Human Recombinant Fetuin A, His Tag
Lieferant: STEMCELL Technologies
Mediate calcium phosphate clearance and prevent ectopic calcification with fetuin A, a plasma glycoprotein that forms soluble complexes with calcium and phosphate (Heiss et al.; Price and Lin). Belonging to the cystatin superfamily of cysteine protease inhibitors (Brown and Dziegielewska), fetuin A has also been shown to play a role in lipid transport, acting as a carrier (Kumbla et al.). In cell-based assays, it has been suggested that fetuin A protects against lethal systemic infection through the inhibition of high mobility group box 1 (HMGB1) protein accumulation and release (Li et al.). Fetuin A acts as a natural antagonist against specific TGF-β and BMP signaling proteins, blocking osteogenic differentiation of rat bone marrow cells (Demetriou et al.). This protein contains a His-residue tag at the carboxyl end of the polypeptide chain. For consistency and reproducibility across your applications, fetuin A from STEMCELL comes lyophilised with ≥94% purity, and endotoxin levels are verified to be ≤1.0 EU/μg protein.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').