Human Recombinant Serpin A12, His tag
Lieferant: STEMCELL Technologies
100-1326
STMC100-1326EA 956 EUR
STMC100-1326
Human Recombinant Serpin A12, His tag
Proteine und Peptide
Commonly known as vaspin, Serpin A12 is an adipokine that is thought to perform insulin-sensitizing functions, such as the regulation of kallikrein 7 (KLK7), an insulin-inhibiting protease (Heiker et al.). The Serpin A12 glycoprotein belongs to the serine protease inhibitor family (serpin), and is mainly associated with obesity, insulin resistance, and glucose metabolism, but may also protect against high fat-induced bone loss (Wang et al.). Serpins share a highly conserved core structure with multiple α-helices, β-strands, and a reactive site loop (RCL) to interact with the target protease (Huntington). It is expressed predominantly in visceral adipose tissue and at significantly higher amounts in obese individuals (Kurowska et al.). This protein contains a His-residue tag at the carboxyl end of the polypeptide chain. For consistency and reproducibility across your applications, Serpin A12 from STEMCELL comes lyophilised with ≥94% purity, and is verified by LAL analysis to ensure endotoxin levels are ≤1,0 EU/μg protein.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').