Human Recombinant Visfatin
Lieferant: STEMCELL Technologies
100-1335
STMC100-1335EA 848 EUR
STMC100-1335
Human Recombinant Visfatin
Proteine und Peptide
Use visfatin to catalyse the production of nicotinamide mononucleotide from nicotinamide (Revollo et al.). Nicotinamide mononucleotide is a precursor to nicotinamide adenine dinucleotide (NAD) that is vital to energy metabolism, cell death, and other cellular processes (Ying). Visfatin also acts as an immunomodulator by activating leukocytes and inducing the production of pro-inflammatory and anti-inflammatory cytokines (Moschen et al.), and there is evidence that visfatin can regulate insulin receptor signaling and insulin secretion (Brown et al.). Belonging to the nicotinate phosphoribosyltransferase (NAPRTase) family, Visfatin is an adipokine that is produced in adipocytes, leukocytes, and hepatocytes (Chiu et al., Garten et al., Kralisch et al.). It is active as a homodimer with each identical monomer consisting of two structural domains made up of 19 β-strands and 13 α-helices (Kim et al.). For consistency and reproducibility across your applications, visfatin from STEMCELL comes lyophilised with ≥90% purity, and is verified by LAL analysis to ensure endotoxin levels are ≤1,0 EU/μg protein.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').