EBV (EBNA-3B) Peptide Pool
Lieferant: STEMCELL Technologies
100-1387
STMC100-1387EA 1040 EUR
STMC100-1387
EBV (EBNA-3B) Peptide Pool
Proteine und Peptide
Stimulating T cells with EBV (EBNA-3B) peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. EBV (EBNA-3B) Peptide Pool is a lyophilised mixture of 234 peptides from Epstein-Barr nuclear antigen 3B (EBNA-3B) of Epstein-Barr virus (EBV; strain AG876) and consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 946 on EBNA-3B. EBNA-3B is considered non-essential for B cell growth transformation (Chen et al., 2005; Tomkinson and Kieff), but has been shown to regulate the expression of cellular genes in EBV-infected cells (Chen at al., 2006). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').