EBV (LMP1) Peptide Pool
Lieferant: STEMCELL Technologies
100-1388
STMC100-1388EA 848 EUR
STMC100-1388
EBV (LMP1) Peptide Pool
Proteine und Peptide
Stimulating T cells with EBV (LMP1) Peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. EBV (LMP1) Peptide pool is a lyophilised mixture of 94 peptides from latent membrane protein 1 (LMP1) of Epstein-Barr virus (EBV; strain B95-8), and consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 386 on LMP1. LMP1 is essential for EBV-mediated growth transformation of B cells (Kaye et al.) and has been shown to downregulate toll-like receptor 9 (TLR9) expression in infected cells, a mechanism the virus may use to escape host innate immune recognition (Fathallah et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').