HHV8 (K8.1) Peptide Pool
Lieferant: STEMCELL Technologies
100-1404
STMC100-1404EA 663 EUR
STMC100-1404
HHV8 (K8.1) Peptide Pool
Proteine und Peptide
Stimulating T cells with HHV8 (K8.1) Peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. HHV8 (K8.1) Peptide pool is a lyophilised mixture of 55 peptides from glycoprotein K8.1 of human herpesvirus 8 (HHV-8) and consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 228 on K8.1. Expressed in the late-lytic phase, K8.1 is a viral envelope protein that binds to heparin sulfate on the cell surface and facilitates HHV 8 infection (Zoeteweij et al; Birkmann et al.; Wang et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').