HHV1 (gD) Peptide Pool
Lieferant: STEMCELL Technologies
100-1406
STMC100-1406EA 509 EUR
STMC100-1406
HHV1 (gD) Peptide Pool
Proteine und Peptide
Stimulating T cells with HHV1 (gD) Peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. HHV1 (gD) Peptide pool is a lyophilised mixture of 96 peptides from envelope glycoprotein D of human herpesvirus 1 (HHV1; strain Patton), and consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 394 on gD. gD is required at the first stage of virus-induced membrane fusion (Subramanian and Geraghty), which likely occurs via binding to receptors such as nectin-1 (Giovine et al.) and HVEM (Connolly et al.) on cell surfaces. Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').