ImmunoCult™ Dendritic Cell Maturation Supplement
Lieferant: STEMCELL Technologies
10989
STMC10989EA 517 EUR
STMC10989
ImmunoCult™ Dendritic Cell Maturation Supplement
Zellkulturmedien
Supplement for the maturation of human monocyte-derived dendritic cells.
Use ImmunoCult™ Dendritic Cell Maturation Supplement for the maturation of immature dendritic cells (DCs) to mature DCs. For your convenience, the medium is available either individually or as part of the ImmunoCult™ Dendritic Cell Culture Kit. The ImmunoCult™ Dendritic Cell Maturation Supplement is formulated for use in combination with ImmunoCult™-ACF Dendritic Cell Medium and ImmunoCult™-ACF Dendritic Cell Differentiation Supplement to support the maturation of immature DCs.
For more information on protocols for maturation of DCs using ImmunoCult™ Dendritic Cell Maturation Supplement, please explorethe Product Information Sheet (PIS).
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').