SpinSep™ Density Medium
Lieferant: STEMCELL Technologies
17531
STMC17531EA 268 EUR
STMC17531
SpinSep™ Density Medium
Zelltrennmedien
Density gradient medium.
  • For use in cell isolation applications
SpinSep™ Density medium is a density gradient medium designed for use with the EasySep™ human myeloid positive selection kit.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').