Immunomagnetic positive selection of human cells labeled with FITC-conjugated antibodies.
- Fast and easy-to-use
- No columns required
The EasySep™ Human FITC Positive Selection Kit II is designed to isolate cells that are labeled with FITC-conjugated antibodies. Desired cells are targeted with antibody complexes recognising FITC and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This product replaces the EasySep™ Human FITC Positive Selection Kit (STMC18552).
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).