EasySep™ APC Positive Selection Kit II
Lieferant: STEMCELL Technologies
17681 17681RF
STMC17681EA 1010 EUR
STMC17681 STMC17681RF
EasySep™ APC Positive Selection Kit II
Zelltrennmedien
Immunomagnetic positive selection of any cells labeled with APC-conjugated antibodies.
  • Fast and easy-to-use
  • No columns required
The EasySep™ APC Positive Selection Kit II is designed to isolate cells that are labeled with APC (allophycocyanin)-conjugated antibodies by positive selection. Desired cells are targeted with antibody complexes recognising APC and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This product replaces the EasySep™ APC Positive Selection Kit (STMC18453) for even faster cell isolations.
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).