EasySep™ Release Human CD3 Positive Selection Kits
Lieferant: STEMCELL Technologies
17751 17751RF
STMC17751EA 1700 EUR
STMC17751 STMC17751RF
EasySep™ Release Human CD3 Positive Selection Kits
Zelltrennmedien
Immunomagnetic positive selection of particle-free human CD3+ cells.
  • Highly purified human CD3+ cells isolated in less than 30 minutes
  • No-wash removal of EasySep™ Releasable RapidSpheres™
Isolate highly purified and magnetic particle-free CD3+ cells from peripheral blood mononuclear cells or washed leukapheresis samples in as little as 30 minutes.
Using immunomagnetic positive selection, CD3+ cells are labeled with antibodies and magnetic particles, then separated without columns using an EasySep™ magnet. Unwanted cells are poured off, while desired cells remain in the tube. Bound magnetic particles are then removed from the isolated CD3+ cells, which are immediately available for downstream use. Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins, or other chemically related ligands. The CD3 antigen is expressed on all T cells and CD56+ NKT cells.
For even higher efficiency, the process can be automated using RoboSep™.
This kit is designed for cell therapy research applications, but may be qualified for use as an ancillary material following the framework outlined in USP. STEMCELL can work with you to qualify this reagent as an ancillary material under an approved investigational new drug.
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).