EasySep™ Human CD271 Positive Selection Kit II
Lieferant: STEMCELL Technologies
17849
STMC17849EA 1420 EUR
STMC17849
EasySep™ Human CD271 Positive Selection Kit II
Zelltrennmedien
Immunomagnetic positive selection of human CD271+ cells.
  • Fast and easy-to-use
  • No columns required
The EasySep™ Human CD271 Positive Selection Kit II is designed to isolate CD271+ cells from fresh human bone marrow by positive selection. Desired cells are targeted with antibody complexes recognising CD271 and dextran-coated magnetic particles. The kit also contains an antibody to human Fc receptor to minimize nonspecific binding. labelled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD271+ antigen is expressed on multiple cell types including mesenchymal stem and progenitor cells (MSCs).
This product replaces the EasySep™ Human CD271 Positive Selection Kit for even faster cell isolations.
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').