Column-free immunomagnetic positive selection of human CD66b+ cells for HLA analysis.
- Fast and easy-to-use
- Up to 99% purity
- No columns required
The EasySep™ HLA Chimerism Whole Blood CD66b Positive Selection Kit is designed to isolate highly purified CD66b+ cells from fresh whole blood by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognising CD66b and dextran-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD66b antigen is expressed on mature granulocytes. Isolated cells are immediately available for downstream applications such as flow cytometry, culture, or DNA/RNA extraction for lineage-specific chimerism analysis.
This product replaces the EasySep™ Human Whole Blood CD66b Positive Selection Kit for even faster cell isolations.
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').