EasySep™ HLA Chimerism Whole Blood CD8 Positive Selection Kits
Lieferant: STEMCELL Technologies
17889 17889RF
STMC17889EA 1460 EUR
STMC17889 STMC17889RF
EasySep™ HLA Chimerism Whole Blood CD8 Positive Selection Kits
Zelltrennmedien
Column-free immunomagnetic positive selection of human CD8+ cells for HLA analysis.
  • Fast and easy-to-use
  • Up to 99,9% purity
  • No columns required
The EasySep™ HLA Chimerism Whole Blood CD8 Positive Selection Kit is designed to isolate CD8+ cells from fresh whole blood or buffy coat by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognising CD8 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. labelled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD8 antigen is expressed on cytotoxic T cells and weakly on a subset of NK cells.
This product replaces the EasySep™ Human Whole Blood CD8 Positive Selection Kit for even faster cell isolations.
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').