EasySep™ Dead Cell Removal (Annexin V) Kit
Lieferant: STEMCELL Technologies
17899
STMC17899EA 578 EUR
STMC17899
EasySep™ Dead Cell Removal (Annexin V) Kit
Zelltrennmedien
Immunomagnetic depletion of apoptotic (Annexin V+) cells from culture or tissue preparations.
  • Fast and easy-to-use
  • No columns required
  • Compatible across EasySep™, 'The Big Easy', and EasyEights™ platforms
The EasySep™ Dead Cell Removal (Annexin V) kit is designed to deplete apoptotic (Annexin V+) cells from cell culture or tissue preparations by immunomagnetic negative selection. Unwanted cells are labelled with Annexin V, antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are simply poured off into a new tube. Annexin V binds phosphatidylserine on the outer leaflet of the cell membrane during apoptosis.
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').