EasySep™ Easy 50 Magnet
Lieferant: STEMCELL Technologies
18002
STMC18002EA 2920 EUR
STMC18002
EasySep™ Easy 50 Magnet
Magnete
The EasySep™ Easy 50 magnet is designed for cell separation procedures using selected EasySep™ reagents when processing a large number of cells (up to 4×10⁹ cells).
  • Immunomagnetic column-free magnet
  • Designed to hold a standard 50 ml conical tube
The EasySep™ Easy 50 magnet generates a high-gradient magnetic field in the interior cavity that is strong enough to separate cells labeled with magnetic EasySep™ particles, without the use of columns.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').