EasySep™ Mouse CD11c Positive Selection Kit II
Lieferant: STEMCELL Technologies
18780 18781RF 18781 18780RF
STMC18780EA 1520 EUR
STMC18780 STMC18781RF STMC18781 STMC18780RF
EasySep™ Mouse CD11c Positive Selection Kit II
Zelltrennmedien
Immunomagnetic positive selection of mouse CD11c+ cells (e.g. dendritic cells (DC)) from mouse splenocytes or other tissues
  • Fast and easy-to-use
  • Up to 95% purity
  • No columns required
  • Isolated cells are not fluorochrome-labelled
The EasySep™ Mouse CD11c Positive Selection Kit II isolates highly purified CD11c+ cells from splenocytes or other tissues by immunomagnetic positive selection. Desired cells are targeted with antibodies and magnetic particles, and isolated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Isolated cells are immediately ready for downstream applications such as flow cytometry, culture, and cell-based experiments.
This product replaces the EasySep™ Mouse CD11c Positive Selection Kit (Catalog #18758) for even faster cell isolations and does not result in the labeling of isolated cells with PE.
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').