EasySep™ Mouse CD25 Regulatory T Cell Positive Selection Kits
Lieferant: STEMCELL Technologies
18782 18782RF
STMC18782EA 1070 EUR
STMC18782 STMC18782RF
EasySep™ Mouse CD25 Regulatory T Cell Positive Selection Kits
Zelltrennmedien
Immunomagnetic positive selection of mouse regulatory T cells.
  • Fast and easy-to-use
  • Up to 93% purity
  • No columns required
The EasySep™ Mouse CD25 Regulatory T Cell Positive Selection Kit is designed to isolate highly purified CD25+ cells from single-cell suspensions of splenocytes or other tissues by positive selection. The kit targets CD25 regulatory T cells for positive selection with a PE-labelled antibody directed against the CD25 surface marker. Desired cells are labelled with antibodies and magnetic particles. The cells are separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube.
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').