EasySep™ Human Pan-B Cell Enrichment Kits
Lieferant: STEMCELL Technologies
19554RF 19554
STMC19554RFEA 2010 EUR
STMC19554RF STMC19554
EasySep™ Human Pan-B Cell Enrichment Kits
Zelltrennmedien
Immunomagnetic negative selection of human pan-B cells
  • Fast, easy-to-use and column-free
  • Up to 99% purity
  • Isolated cells are untouched
The EasySep™ Human Pan-B Cell Enrichment Kit is designed to isolate B cells including plasma cells from normal fresh or previously frozen peripheral blood mononuclear cells by negative selection. Undesired cells are targeted for removal with Tetrameric Antibody Complexes recognising CD2, CD3, CD14, CD16, CD36, CD42b, CD56, CD66b, CD123, glycophorin A and dextran-coated magnetic particles. The labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').