EasySep™ Rat Custom Enrichment Kits
Lieferant: STEMCELL Technologies
19609 19609RF
STMC19609EA 0 EUR
STMC19609 STMC19609RF
EasySep™ Rat Custom Enrichment Kits
Zelltrennmedien
Immunomagnetic negative selection of rat cells
The EasySep™ Rat Custom Enrichment Kit is designed to isolate any cell type of interest from a single-cell suspension by negative selection. STEMCELL Technologies offers a wide selection of antibodies for custom separation and will work with you to isolate a cell type of interest. Please contact our technical support team for more information.
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').