EasySep™ Direct Human Pan-Granulocyte Isolation Kit
Lieferant: STEMCELL Technologies
19659
STMC19659EA 1100 EUR
STMC19659
EasySep™ Direct Human Pan-Granulocyte Isolation Kit
Zelltrennmedien
Immunomagnetic isolation of human granulocytes directly from whole blood
  • >99,9% RBC depletion without the need for density gradient centrifugation, sedimentation or lysis
  • Up to 99% purity of isolated cells
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
The EasySep™ Direct Human Pan-Granulocyte Isolation Kit isolates functional, highly purified neutrophils, eosinophils, and basophils directly from human whole blood by immunomagnetic negative selection. No lysis, density gradient centrifugation, or other processing steps are required and isolate​ed cells are immediately available for flow cytometry, functional assays, culture, and other downstream applications.
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').