EasySep™ Mouse Pan-DC Enrichment Kits
Lieferant: STEMCELL Technologies
19763 19763RF
STMC19763EA 1580 EUR
STMC19763 STMC19763RF
EasySep™ Mouse Pan-DC Enrichment Kits
Zelltrennmedien
Immunomagnetic isolation of untouched mouse pan-dendritic cells (DCs)
  • Fast, easy-to-use and column-free
  • Up to 76% purity
  • Isolated cells are untouched
The EasySep™ Mouse Pan-DC Enrichment Kit is designed to isolate pan-dendritic cells which isolates cells in just 13 minutes.
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').