EasySep™ Mouse ILC2 Enrichment Kit
Lieferant: STEMCELL Technologies
19842
STMC19842EA 1640 EUR
STMC19842
EasySep™ Mouse ILC2 Enrichment Kit
Zelltrennmedien
Immunomagnetic enrichment of untouched mouse ILC2 cells
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
  • Facilitates rapid flow sorting of ILC2s
This product is designed to enrich ILC2's (Group 2 Innate Lymphoid Cells) from single-cell suspensions of lung tissue or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-ILC2 cells and streptavidin-coated magnetic particles (RapidSpheres™). labelled cells are separated using an EasySep™ magnet without the use ofMore Product Information