EasySep™ Mouse ILC2 Enrichment Kit
Lieferant: STEMCELL Technologies
19842
STMC19842EA 1640 EUR
STMC19842
EasySep™ Mouse ILC2 Enrichment Kit
Zelltrennmedien
Immunomagnetic enrichment of untouched mouse ILC2 cells
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
  • Facilitates rapid flow sorting of ILC2s
This product is designed to enrich ILC2's (Group 2 Innate Lymphoid Cells) from single-cell suspensions of lung tissue or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-ILC2 cells and streptavidin-coated magnetic particles (RapidSpheres™). labelled cells are separated using an EasySep™ magnet without the use of columns.
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').